Learn More
                                    Mölnlycke™ Biogel™ NeoTech Gloves
Sterile cleanroom gloves with beaded cuffs and curved fingers are suitable for use in aseptic and Class 100 (ISO 5) / EU GMP Grade A environments. Adviced to be worn in combination with the Biogel™ NeoTech Indicator™ Underglove to form a breach indication system.
Marque: Mölnlycke™ 4450990-02
                                Les retours ne sont pas autorisés pour ce produit.
                                Afficher la politique du retour.
                Description
- Can be worn together with Biogel™ NeoTech Indicator™ Underglove to form a breach indication system
 - Tested and cleared for use with chemotherapy agents
 - Synthetic polychloroprene
 - Biogel hydrogel polymer coating for easy donning
 - Exceed industry standard for force at break, tensile strength and elongation
 - Every glove (100%) is air-inflation tested for holes typically not detected in a visual inspection
            Spécification
| ASTM D3577 Viral penetration ASTM F1671 Pyrogenicity ASTM D7102 | |
| Class 100 (ISO 5) | |
| 0.65 | |
| Single-use | |
| 8,3 mil | |
| Spécifique | |
| Polychloroprène | |
| 0.19 mm | |
| 9 | |
| 6.7 mil (Cuff) | |
| 21 MPa | |
| 30 cm | 
| EN 455, ISO 10282 , EN 374, EN 16523-1, PPE Cat. III, Quality/Environment ISO 13485, ISO 14001, Sterilisation ISO 11137, Viral penetration ISO 16604, Allergenicity ISO 10993, Labelling EN 556-1, EN ISO 15223-1, EN ISO 21420 | |
| No | |
| Blanc | |
| Lisse | |
| 0.21 mm | |
| Oui | |
| 7.5 mil | |
| Sans poudre | |
| Stérile | |
| 0.17 mm | |
| 11,8 in | 